MilliporeSigma
  • Vitamin D reduces the differentiation and expansion of Th17 cells in young asthmatic children.
Vitamin D reduces the differentiation and expansion of Th17 cells in young asthmatic children.
Immunobiology (2014-08-17)
Agnes Hamzaoui, Anissa Berraïes, Besma Hamdi, Wajih Kaabachi, Jamel Ammar, Kamel Hamzaoui
ABSTRACT
Vitamin D [25(OH)D3] deficiency has been associated with asthma as in many inflammatory and autoimmune pathologies; however, there is still a lack of data about the effects of administration of vitamin D in immune regulation in young asthmatic patients. In this study, we investigated its inhibitory effect on the immune response in young asthmatic patients and the possible mechanisms involved. Peripheral blood CD4(+) T cells from 10 asthmatic patients and 10 healthy controls were cultured under Th17 polarizing conditions in the presence or absence of [25(OH)D3], IL-17 cytokine production was determined by ELISA and flow cytometry. Messenger RNA (mRNA) expression of several factors related to Th17 cell function was determined by real-time PCR. The effect of [25(OH)D3]-treated dendritic cells (DCs) on CD4(+) T cell response was determined by ELISA and flow cytometry. Stimulation of naive CD4(+) T cells under Th17 polarizing conditions showed a higher Th17 cell differentiation in asthmatic patients than healthy controls. The addition of [25(OH)D3] significantly inhibited Th17 cell differentiation both in patients [P<0.001] and in normal controls [P=0.001] in a dose-dependent way. [25(OH)D3] was able to inhibit the gene expression of RORC, IL-17, IL-23R, and CCR6. [25(OH)D3]-treated DCs significantly inhibited IL-17 production [P=0.002] and decreased the percentage of CD4(+)IL-17(+) [P=0.007] in young asthmatics. The findings suggest that the inhibitory effect of [25(OH)D3] on the Th17 response was mediated via both T cells and DCs. DCs pathway is involved in the direct inhibition of 25(OH)D3 on Th17 cell differentiation in young asthmatics.
Sigma-Aldrich
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Human IL-17 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Mouse IL-17 (IL-17A) ELISA Kit, for serum, plasma and cell culture supernatant