MilliporeSigma
  • Microsphaerol and seimatorone: two new compounds isolated from the endophytic fungi, Microsphaeropsis sp. and Seimatosporium sp.
Microsphaerol and seimatorone: two new compounds isolated from the endophytic fungi, Microsphaeropsis sp. and Seimatosporium sp.
Chemistry & biodiversity (2015-02-14)
Hidayat Hussain, Natalia Root, Farah Jabeen, Ahmed Al-Harrasi, Manzoor Ahmad, Fazal Mabood, Zahid Hassan, Afzal Shah, Ivan R Green, Barbara Schulz, Karsten Krohn
ABSTRACT
A new polychlorinated triphenyl diether named microsphaerol (1), has been isolated from the endophtic fungus Microsphaeropsis sp. An intensive phytochemical investigation of the endophytic fungus Seimatosporium sp., led to the isolation of a new naphthalene derivative named seimatorone (2) and eight known compounds, i.e., 1-(2,6-dihydroxyphenyl)-3-hydroxybutan-1-one (3), 1-(2,6-dihydroxyphenyl)butan-1-one (4), 1-(2-hydroxy-6-methoxyphenyl)butan-1-one (5), 5-hydroxy-2-methyl-4H-chromen-4-one (6), 2,3-dihydro-5-hydroxy-2-methyl-4H-chromen-4-one (7), 8-methoxynaphthalen-1-ol (8), nodulisporins A and B (9 and 10, resp.), and daldinol (11). The structures of 1 and 2 were elucidated by detailed spectroscopic analysis including (1) H- and (13) C-NMR, COSY, HMQC, HMBC, and HR-EI-MS, while the structures of the known compounds were deduced from comparison of their spectral data with those in the literature. Preliminary studies revealed that microsphaerol (1) showed good antibacterial activities against B. Megaterium and E. coli, and good antilagal and antifungal activities against C. fusca, M. violaceum, respectively. On the other hand, seimatorone (2) exhibited moderate antibacterial, antialgal, and antifungal activities.
Sigma-Aldrich
Cerium(IV) sulfate