MilliporeSigma
  • Self‑assembled monolayers of alkanethiolates on surface chemistry groups in osteosarcoma cells.
Self‑assembled monolayers of alkanethiolates on surface chemistry groups in osteosarcoma cells.
Molecular medicine reports (2014-11-07)
Ying-Hu Deng, Li-Hua Li, Jin He, Mei Li, Yu Zhang, Xiu-Mei Wang, Fu-Zhai Cui, Hong Xia
ABSTRACT
Cell biomedical behavior is influenced by a number of factors, and the extracellular matrix (ECM) of the cellular microenvironment affects certain cancer cells. In the current study, U‑2OS cells were cultured on gold surfaces modified with different terminal chemical groups [methyl (‑CH3), amino (‑NH2), hydroxyl (‑OH) and carboxyl (‑COOH)]. The results revealed that different chemical surfaces convey different behaviors. The density of the different functional surfaces was confirmed by atomic force microscopy. Cell morphology, proliferation rate and cell cycle were investigated using scanning electron microscopy, cell counting and flow cytometry. In conclusion, the type of chemical group on a biomaterial is an important property for the growth of osteosarcoma cells; ‑NH2 and ‑COOH surfaces sustained visible cell adhesion and promoted cell growth.
Sigma-Aldrich
11-Mercapto-1-undecanol, 97%
Sigma-Aldrich
1-Undecanethiol, 98%
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
11-Mercapto-1-undecanol, 99%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
12-Mercaptododecanoic acid, 96%
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Supelco
Dimethyl sulfoxide, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications