MilliporeSigma
  • Inhibition of tissue factor:factor VIIa-catalyzed factor IX and factor X activation by TFPI and TFPI constructs.
Inhibition of tissue factor:factor VIIa-catalyzed factor IX and factor X activation by TFPI and TFPI constructs.
Journal of thrombosis and haemostasis : JTH (2014-08-29)
S Peraramelli, S Thomassen, A Heinzmann, J Rosing, T M Hackeng, R Hartmann, F Scheiflinger, M Dockal
ABSTRACT
TFPI is a Kunitz-type protease inhibitor that downregulates the extrinsic coagulation pathway by inhibiting factor Xa (FXa) and FVIIa. All three Kunitz domains (KD1, KD2, and KD3) and protein S are required for optimal inhibition of FXa and FVIIa. There is limited information on Kunitz domain requirements of the inhibition of TF:FVIIa-catalyzed FIX and FX activation by TFPI. To investigate the role of the Kunitz domains of TFPI and protein S in the inhibition of FX and FIX activation. Inhibition of TF:FVIIa-catalyzed FX and FIX activation by full-length TFPI (TFPIFL ) and TFPI constructs was quantified from progress curves of FXa and FIXa generation measured with chromogenic substrates. TFPIFL inhibited TF:FVIIa-catalyzed FIX activation with a Ki of 16.7 nmol L(-1) . Protein S reduced the Ki to 1.0 nmol L(-1) . TFPI1-150 and KD1-KD2 had 10-fold higher Ki values and were not stimulated by protein S. Single Kunitz domains were poor inhibitors of TF:FVIIa-catalyzed FIX activation (Ki >800 nm). FX activation was measured at limiting FVIIa and excess TF or vice versa. At both conditions, TFPIFL , TFPI1-150 , and KD1-KD2 showed similar inhibition of FX activation. However, at low phospholipid concentrations, TFPIFL was ~ 15-fold more active than TFPI1-150 or KD1-KD2. Apparently, excess phospholipids act as a kind of sink for TFPIFL , limiting its availability for TF:FVIIa inhibition. Preformed FXa:TFPIFL/1-150 complexes rapidly and stoichiometrically inhibited FIX and FX activation by TF:FVIIa, indicating that binary TFPI:FXa complex formation is the limiting step in TF:FVIIa inhibition. Protein S also enhanced inhibition of TF:FVIIa-catalyzed FX activation by TFPI.
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥97.0% (TLC)
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥99.0% (10 mg phospholipid per ml CHCl3, TLC)