MilliporeSigma
  • Efficient acid-catalyzed (18) F/(19) F fluoride exchange of BODIPY dyes.
Efficient acid-catalyzed (18) F/(19) F fluoride exchange of BODIPY dyes.
ChemMedChem (2014-03-07)
Edmund J Keliher, Jenna A Klubnick, Thomas Reiner, Ralph Mazitschek, Ralph Weissleder
ABSTRACT
Fluorine-containing fluorochromes are important validation agents for positron emission tomography imaging compounds, as they can be readily validated in cells by fluorescence imaging. In particular, the (18) F-labeled BODIPY-FL fluorophore has emerged as an important platform, but little is known about alternative (18) F-labeling strategies or labeling on red-shifted fluorophores. In this study we explore acid-catalyzed (18) F/(19) F exchange on a range of commercially available N-hydroxysuccinimidyl ester and maleimide BODIPY fluorophores. We show this method to be a simple and efficient (18) F-labeling strategy for a diverse span of fluorescent compounds, including a BODIPY-modified PARP-1 inhibitor, and amine- and thiol-reactive BODIPY fluorophores.
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%