MilliporeSigma
  • Influence of alkalizers on dissolution properties of telmisartan in solid dispersions prepared by cogrinding.
Influence of alkalizers on dissolution properties of telmisartan in solid dispersions prepared by cogrinding.
Drug development and industrial pharmacy (2013-10-09)
Lin Zhong, Xingyi Zhu, Bo Yu, Weike Su
ABSTRACT
The amorphous solid dispersions of telmisartan salts were prepared by cogrinding, in presence of alkalizers and polyvinylpyrrolidone (PVPk30). Five alkalizers in this study were MgO, Na2CO3, K2CO3, NaHCO3 and meglumine. In soft mode using a roll mill, the drug could not form salt with MgO or NaHCO3, whereas partial drug had been transformed into salt with carbonates or meglumine. Under cogrinding, the organic base meglumine was easier to react with telmisartan than other two carbonates. For getting good dissolution performance, the drug had to be transformed into salt completely. A high intensity oscillating mill was applied for producing telmisartan meglumine salt. Multi-instrumental characterizations attested the formation of amorphous salt by high mechanical process, involving dissolution test, fourier transform infrared spectroscopy and powder X-ray diffractometry. It was evident that solid dispersions of telmisartan meglumine salt significantly increased the drug dissolution rate in intestinal fluid.
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)