MilliporeSigma
  • Combined effect of pH and high temperature on the stability and antioxidant capacity of two anthocyanins in aqueous solution.
Combined effect of pH and high temperature on the stability and antioxidant capacity of two anthocyanins in aqueous solution.
Food chemistry (2014-06-11)
Xiaonan Sui, Xin Dong, Weibiao Zhou
ABSTRACT
The stability of two cyanidin-based anthocyanins from black rice in an aqueous system containing them with a pH range of 2.2-6.0 was investigated at temperatures ranging from 100 to 165°C. Within these pH and temperature ranges, the stability of the two anthocyanins was found to decrease gradually with increasing pH. The lowest (8.99×10(-4)s(-1)) and highest (0.120s(-1)) degradation rate constants for cyanidin-3-glucoside were obtained at pH 2.2 & 100°C and pH 6.0 & 165°C, respectively, whereas those for cyanidin-3-rutinoside were 5.33×10(-4)s(-1) at pH 2.2 & 100°C and 7.39×10(-2)s(-1) at pH 5.0 & 165°C, respectively. Antioxidant capacity analysis was conducted on thermally processed anthocyanin solutions to further evaluate the effects of pH and thermal treatment on the anthocyanins. The total antioxidant capacity of samples after thermal treatments under various pHs was found to remain at a similar level.
Product Description
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri