MilliporeSigma
  • Removal of Pb(II) ions from aqueous solution using water hyacinth root by fixed-bed column and ANN modeling.
Removal of Pb(II) ions from aqueous solution using water hyacinth root by fixed-bed column and ANN modeling.
Journal of hazardous materials (2014-04-15)
Tania Mitra, Biswajit Singha, Nirjhar Bar, Sudip Kumar Das
ABSTRACT
Hyacinth root was used as a biosorbent for generating adsorption data in fixed-bed glass column. The influence of different operating parameters like inlet Pb(II) ion concentration, liquid flow rate and bed height on the breakthrough curves and the performance of the column was studied. The result showed that the adsorption efficiency increased with increase in bed height and decreased with increase in inlet Pb(II) ion concentration and flow rate. Increasing the flow rate resulted in shorter time for bed saturation. The result showed that as the bed height increased the availability of more number of adsorption sites in the bed increased, hence the throughput volume of the aqueous solution also increased. The adsorption kinetics was analyzed using different models. It was observed that maximum adsorption capacity increased with increase in flow rate and initial Pb(II) ion concentration but decreased with increase in bed height. Applicability of artificial neural network (ANN) modeling for the prediction of Pb(II) ion removal was also reported by using multilayer perceptron with backpropagation, Levenberg-Marquardt and scaled conjugate algorithms and four different transfer functions in a hidden layer and a linear output transfer function.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%