MilliporeSigma
  • Experimental study of survival of pedicled perforator flap with flow-through and flow-end blood supply.
Experimental study of survival of pedicled perforator flap with flow-through and flow-end blood supply.
The British journal of surgery (2015-02-18)
Y Wang, S-Y Chen, W-Y Gao, J Ding, W Shi, X-L Feng, X-Y Tao, L Wang, D-S Ling
ABSTRACT
Flap viability after transfer depends on blood flow from the arterial blood supply below the fascia. This study evaluated survival of a pedicle flap with a perforator lateral branch and flow-through blood supply, compared with that of a flap with a flow-end blood supply and perforator terminal branch. Forty Sprague-Dawley rats, 20 in each group, were assigned to transfer of a superficial epigastric artery pedicle island flap with a flow-through or flow-end configuration of blood supply. Laser Doppler imaging was used to evaluate flap perfusion 2 h, 3 days and 5 days after surgery. The rats were killed on day 5, and lead oxide-gelatine-enhanced flap angiography and histology with haematoxylin and eosin staining was performed. Dorsal midline tissue was excised for quantification of vascular endothelial growth factor by western blot assay. On day 5 after surgery, the flow-through group exhibited a significantly greater mean(s.d.) flap survival area (97·8(3·5) versus 80·8(10·2) per cent; P = 0·003), microvascular density (303(19) versus 207(41) per mm(2) ; P < 0·001) and perfusion (8·64(0·14) versus 5·95(0·14) perfusion units; P < 0·001) than the flow-end group. The flow-through group exhibited more angiosomes connected by dilated vascular anastomoses between the skin and subcutaneous fasciae. The flow-through blood supply improved pedicle perforator flap survival. Surgical relevance Perforator flap failure is mainly the result of impaired blood supply, as a flow-end blood configuration is nourished only by the perforator terminal branch of the artery. This work showed that the flow-through blood supply nourished by the perforator lateral branch improved flap survival, with dilatation of collateral vascular anastomoses and increased neoangiogenesis. The use of a flow-through configuration improves perforator flap survival and could therefore minimize morbidity resulting from flap necrosis.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture