MilliporeSigma
  • Prolongation of overall survival in advanced lung adenocarcinoma patients with the XAGE1 (GAGED2a) antibody.
Prolongation of overall survival in advanced lung adenocarcinoma patients with the XAGE1 (GAGED2a) antibody.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-08-16)
Yoshihiro Ohue, Koji Kurose, Yu Mizote, Hirofumi Matsumoto, Yumi Nishio, Midori Isobe, Minoru Fukuda, Akiko Uenaka, Mikio Oka, Eiichi Nakayama
ABSTRACT
The cancer/testis antigen XAGE1 (GAGED2a) is expressed in approximately 40% of advanced lung adenocarcinomas. We investigated the clinical relevance of the XAGE1 (GAGED2a) immune responses in patients with advanced lung adenocarcinoma. The XAGE1 (GAGED2a) antigen expression and EGFR mutation were determined with tumor tissues. The XAGE1 (GAGED2a) antibody and T-cell immune responses, as well as immune cell phenotypes, were analyzed with blood samples. Patients with EGFR wild-type (EGFRwt) tumors were treated with conventional platinum-based doublet chemotherapy and patients with EGFR-mutated (EGFRmt) tumors were treated with EGFR-TKI and conventional chemotherapy. The overall survival (OS) rates of the antibody-positive and -negative patients were investigated. The results showed that the OS of antibody-positive patients was prolonged significantly compared with that of antibody-negative patients with either XAGE1 (GAGED2a) antigen-positive EGFRwt (31.5 vs. 15.6 months, P = 0.05) or EGFRmt (34.7 vs. 11.1 months, P = 0.001) tumors. Multivariate analysis showed that the presence of the XAGE1 (GAGED2a) antibody was a strong predictor for prolonged OS in patients with XAGE1 (GAGED2a) antigen-positive tumors and in patients with either EGFRwt or EGFRmt tumors. On the other hand, XAGE1 (GAGED2a) antigen expression was a worse predictor in patients with EGFRmt tumors. Phenotypic and functional analyses of T cells indicated immune activation in the antibody-positive patients. The findings suggest that production of the XAGE1 (GAGED2a) antibody predicts good prognosis for patients with lung adenocarcinoma as an immune biomarker and the protective effect of this naturally occurring immune response supports the concept of immunotherapy.
Sigma-Aldrich
Interleukin-7 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-7 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-7 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Anti-CD244 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
IL-7 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture