MilliporeSigma
  • Design, synthesis and evaluation of 3,4-dihydroxybenzoic acid derivatives as antioxidants, bio-metal chelating agents and acetylcholinesterase inhibitors.
Design, synthesis and evaluation of 3,4-dihydroxybenzoic acid derivatives as antioxidants, bio-metal chelating agents and acetylcholinesterase inhibitors.
Journal of enzyme inhibition and medicinal chemistry (2014-02-13)
Laura Friggeri, Daniela De Vita, Fabiana Pandolfi, Silvano Tortorella, Roberta Costi, Roberto Di Santo, Luigi Scipione
ABSTRACT
Metal ions, especially copper, zinc and iron, play an important role in the neurodegeneration process because they can affect protein misfolding, leading to the formation of the amyloid deposits and oxidative stress leading to reactive oxygen species (ROS). Here we report the synthesis and evaluation as antioxidant and metal chelating agents of 3,4-dihydroxybenzoic acid derivatives. Synthesized compounds were tested by the 2,2-diphenyl-2-picrylhydrazyl (DPPH) method showing a radical scavenging ability (EC50=0.093-0.118 μM) higher than Trolox used as reference. Furthermore, these compounds were able to bind both iron and copper, especially the iron (III), by the formation of hexa-coordinated complexes. Synthesized compounds were tested to evaluate their ability to inhibit acetyl- and butyryl-cholinesterase; the obtained results have demonstrated that they are selective inhibitors of AChE (Ki=1.5-18.9 μM) and result weakly active versus butyrylcholinesterase (BChE).
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Butyrylthiocholine iodide, ≥98%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Butyrylthiocholine iodide, puriss., ≥99.0% (AT)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
1,3-Propylene sulfite, 99%