MilliporeSigma
  • Structural evaluation of tandem hairpin pyrrole-imidazole polyamides recognizing human telomeres.
Structural evaluation of tandem hairpin pyrrole-imidazole polyamides recognizing human telomeres.
Journal of the American Chemical Society (2014-07-19)
Akiyoshi Hirata, Kiyoshi Nokihara, Yusuke Kawamoto, Toshikazu Bando, Asuka Sasaki, Satoru Ide, Kazuhiro Maeshima, Takeshi Kasama, Hiroshi Sugiyama
ABSTRACT
A polyamide containing N-methylpyrrole (Py) and N-methylimidazole (Im), designated PIPA, binds with high affinity and specificity to specific nucleotide sequences in the minor groove of double-helical DNA. Based on a recent report of the synthesis of PIPA for telomere visualization, the present paper focused on the size of the connecting part (hinge region) of two PIPA segments of the tandem hairpin PIPA, Dab(Im-Im-Py)-Py-Py-Py-Im-[Hinge]-Dab(Im-Im-Py)-Py-Py-Py-Im-βAla-NH(CH2)3N(CH3)-(CH2)3NH-[Dye]. The present paper also describes the characterization of binding by measuring the thermal melting temperature and surface plasmon resonance and by specific staining of telomeres (TTAGGG)n in human cells. Microheterogeneity was also investigated by high-resolution mass spectrometry. We found that the optimal compound as the hinge segment for telomere staining was [-NH(C2H4O)2(C2H4)CO-] with tetramethylrhodamine as the fluorescent dye.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
HEPES
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)