MilliporeSigma
  • Enhancement of catecholamine release from PC12 cells by the traditional Japanese medicine, rikkunshito.
Enhancement of catecholamine release from PC12 cells by the traditional Japanese medicine, rikkunshito.
BMC complementary and alternative medicine (2014-07-21)
Yuko Nagamura, Kiyoshi Terawaki, Yasuhito Uezono, Toshihiko Tsukada
ABSTRACT
Rikkunshito is a traditional Japanese herbal medicine that is used to treat appetite loss associated with cancer and other disorders. The formulation contains various constituents that influence cell signaling, and rikkunshito may accordingly affect human homeostasis through multiple regulatory pathways, including those governed by the endocrine system. We investigated the actions of rikkunshito on catecholamine release from PC12 cells, an adrenal chromaffin cell line. The actions of rikkunshito on PC12 cells were evaluated by measuring intracellular cAMP levels, tyrosine hydroxylase (TH) and vasoactive intestinal peptide (VIP) mRNA expression levels, and catecholamine levels in the culture medium. The transcriptional activation of VIP gene by rikkunshito was assessed by using a VIP promoter-driven reporter gene assay. Rikkunshito dose-dependently enhanced forskolin-induced elevations in cAMP in PC12 cells, and also increased the gene expression of TH and VIP. The transcriptional activation of VIP gene by rikkunshito was confirmed. Norepinephrine and dopamine secretion into the culture medium of PC12 cells were also dose-dependently augmented by rikkunshito and/or forskolin, but experiments with a protein kinase C (PKC) activator and a phosphodiesterase inhibitor revealed that the effects of rikkunshito were not simply due to the modulation of PKC or phosphodiesterase activity. These findings suggest that rikkunshito enhances the release of catecholamines by a novel mechanism involving cAMP.
Product Description
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Supelco
Oleic acid, Selectophore, ≥99%
Oleic acid, European Pharmacopoeia (EP) Reference Standard