MilliporeSigma
  • Identification and characterization of genes, encoding the 3-hydroxybutyrate dehydrogenase and a putative lipase, in an avirulent spontaneous Legionella pneumophila serogroup 6 mutant.
Identification and characterization of genes, encoding the 3-hydroxybutyrate dehydrogenase and a putative lipase, in an avirulent spontaneous Legionella pneumophila serogroup 6 mutant.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2015-01-06)
Maria Scaturro, Cristina Barello, Melania De Giusti, Stefano Fontana, Federica Pinci, Maria Gabriella Giuffrida, Maria Luisa Ricci
ABSTRACT
Legionella pneumophila is a pathogen widespread in aquatic environment, able to multiply both within amoebae and human macrophages. The aim of this study was to identify genes differently expressed in a spontaneous avirulent Legionella pneumophila serogroup 6 mutant, named Vir-, respect the parental strain (Vir+), and to determine their role in the loss of virulence. Protein profiles revealed some differences in Vir- proteomic maps, and among the identified proteins the undetectable 3-hydroxybutyrate dehydrogenase (BdhA) and a down-produced lipase. Both Legionella enzymes were studied before and were here further characterized at genetic level. A significant down-regulation of both genes was observed in Vir- at the transcriptional level, but the use of defined mutants demonstrated that they did not affect the intracellular multiplication. A mutant (MS1) showed an accumulation of poly-3-hydroxybutyrate (PHB) granules suggesting a role of bdhA gene in its degradation process. The lipase deduced amino acid sequence revealed a catalytic triad, typical of the 'lipase box' characteristic of PHB de-polymerase enzymes, that let us suppose a possible involvement of lipase in the PHB granule degradation process. Our results revealed unexpected alterations in secondary metabolic pathways possibly linking the loss of virulence to Legionella lack of energy sources.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thymidine, ≥99%
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture