MilliporeSigma
  • Polyethylenimine derivate conjugated with RGD-TAT-NLS as a novel gene vector.
Polyethylenimine derivate conjugated with RGD-TAT-NLS as a novel gene vector.
Bio-medical materials and engineering (2014-09-18)
Wenfang Zhao, Kehai Liu, Shunsheng Chen, Man Man Zhu, Hui Lv, Jing Hu, Yuan Mao
ABSTRACT
To solve the contradiction between the cell toxicity and transfection efficiency of polyethylenimine (PEI) derivate in non-viral gene therapy, a novel gene vector, P123-PEI-R18 was synthesized by using biodegradable PEI derivate conjugated with trifunctional peptide RGD-TAT-NLS. The particle size of P123-PEI-R18/DNA was around 100-250 nm. The gene vector could condense DNA at the weight ratio of 2 and protect plasmid DNA from being dissolved in the blood circulation. Importantly, the complexes exhibited lower cell toxicity and higher transfection efficiency contrasted with PEI 25 kDa in vitro. P123-PEI-R18 holds high potential as a safe and efficient gene vector.
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Rhein, technical grade
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Rhein
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Triethylamine, analytical standard
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Supelco
Benzene, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Triphosgene, reagent grade, 98%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG