MilliporeSigma
  • Protective effect of Xin Mai Jia ultrafiltration extract on human umbilical vein endothelial cell injury induced by hydrogen peroxide and the effect on the NO-cGMP signaling pathway.
Protective effect of Xin Mai Jia ultrafiltration extract on human umbilical vein endothelial cell injury induced by hydrogen peroxide and the effect on the NO-cGMP signaling pathway.
Experimental and therapeutic medicine (2014-06-20)
Yaling Yin, Jia Wan, Peng Li, Yanlong Jia, Ruili Sun, Guopin Pan, Guangrui Wan
ABSTRACT
The aim of the present study was to evaluate the protective effect of the ultrafiltration extract of Xin Mai Jia (XMJ) on a human umbilical vein endothelial cell (HUVEC) injury model induced by hydrogen peroxide (H
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl carbonate, 99%
Sigma-Aldrich
Diethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
USP
Lovastatin, United States Pharmacopeia (USP) Reference Standard
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Lovastatin, European Pharmacopoeia (EP) Reference Standard