MilliporeSigma
  • Stability studies of antiandrogenic compounds in Curcuma aeruginosa Roxb. extract.
Stability studies of antiandrogenic compounds in Curcuma aeruginosa Roxb. extract.
The Journal of pharmacy and pharmacology (2014-06-07)
Nungruthai Suphrom, Jukkarin Srivilai, Ganniga Pumthong, Nantaka Khorana, Neti Waranuch, Nanteetip Limpeanchob, Kornkanok Ingkaninan
ABSTRACT
Curcuma aeruginosa Roxb. extract is a 5α-reductase antagonist that can be used to treat hair loss. We aimed to study the stability of antiandrogenic constituents, germacrone and other sesquiterpene components in the extract. Germacrone and the extract were analyzed as solid forms or solublized with polyethylene glycol-40 (PEG-40) or methanol using high-performance liquid chromatography and gas chromatography with flame ionization detector. The effects of pH, temperature and light on their stability were studied. Degradation of antiandrogenic compounds in C. aeruginosa was highly sensitive to temperature especially pure anhydrous germacrone, which was completely lost within 3 days at 45°C. Curiously, degradation was slower than as a dried extract. Paradoxically, when solubilized with PEG-40, it was largely intact even after 90 days at 45°C. The MS spectrum of a major degradation product suggested that it was elemenone probably produced by Cope rearrangement. Two other putative degradation products were germacrone-1,10-epoxide and germacrone-4,5-epoxide suggesting that oxidation of double bonds was an important mechanism. Germacrone stability was unaffected by pH (2.0-9.0) but only as dried extract it was slightly degraded by light. Antiandrogenic constituents of C. aeruginosa were instable at high temperature and in solid form. Thus, the extract would be optimately stored as a solution or otherwise as solid form at low temperature.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%