MilliporeSigma
  • CE-TOF MS-based metabolomic profiling revealed characteristic metabolic pathways in postmortem porcine fast and slow type muscles.
CE-TOF MS-based metabolomic profiling revealed characteristic metabolic pathways in postmortem porcine fast and slow type muscles.
Meat science (2014-08-12)
Susumu Muroya, Mika Oe, Ikuyo Nakajima, Koichi Ojima, Koichi Chikuni
ABSTRACT
To determine key compounds and metabolic pathways associated with meat quality, we profiled metabolites in postmortem porcine longissimus lumborum (LL) and vastus intermedius (VI) muscles with different aging times by global metabolomics using capillary electrophoresis-time of flight mass spectrometry. Loading analyses of the principal component analysis showed that hydrophilic amino acids and β-alanine-related compounds contributed to the muscle type positively and negatively, respectively, whereas glycolytic and ATP degradation products contributed to aging time. At 168h postmortem, LL samples were characterized by abundance of combinations of amino acids, dipeptides, and glycolytic products, whereas the VI samples were characterized by abundance of both sulfur-containing compounds and amino acids. The AMP and inosine contents in the VI were approx. 10 times higher than those in the LL at 4h postmortem, suggesting different rates of inosine 5'-monophosphate (IMP) accumulation by adenylate kinase 7 and 5'-nucleotidase, and subsequent different production levels of IMP and hypoxanthine between these two porcine muscles.
Product Description
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
USP
L-Leucine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, ≥99.5%