MilliporeSigma
  • Mogamulizumab, an anti-CCR4 antibody, targets human T-lymphotropic virus type 1-infected CD8+ and CD4+ T cells to treat associated myelopathy.
Mogamulizumab, an anti-CCR4 antibody, targets human T-lymphotropic virus type 1-infected CD8+ and CD4+ T cells to treat associated myelopathy.
The Journal of infectious diseases (2014-08-12)
Junji Yamauchi, Ariella Coler-Reilly, Tomoo Sato, Natsumi Araya, Naoko Yagishita, Hitoshi Ando, Yasuo Kunitomo, Katsunori Takahashi, Yuetsu Tanaka, Yugo Shibagaki, Kusuki Nishioka, Toshihiro Nakajima, Yasuhiro Hasegawa, Atae Utsunomiya, Kenjiro Kimura, Yoshihisa Yamano
ABSTRACT
Human T-lymphotropic virus type 1 (HTLV-1) can cause chronic spinal cord inflammation, known as HTLV-1-associated myelopathy/tropical spastic paraparesis (HAM/TSP). Since CD4(+)CCR4(+) T cells are the main HTLV-1 reservoir, we evaluated the defucosylated humanized anti-CCR4 antibody mogamulizumab as a treatment for HAM/TSP. We assessed the effects of mogamulizumab on peripheral blood mononuclear cells from 11 patients with HAM/TSP. We also studied how CD8(+) T cells, namely CD8(+) CCR4(+) T cells and cytotoxic T lymphocytes, are involved in HTLV-1 infection and HAM/TSP pathogenesis and how they would be affected by mogamulizumab. Mogamulizumab effectively reduced the HTLV-1 proviral load (56.4% mean reduction at a minimum effective concentration of 0.01 µg/mL), spontaneous proliferation, and production of proinflammatory cytokines, including interferon γ (IFN-γ). Like CD4(+)CCR4(+) T cells, CD8(+)CCR4(+) T cells from patients with HAM/TSP exhibited high proviral loads and spontaneous IFN-γ production, unlike their CCR4(-) counterparts. CD8(+)CCR4(+) T cells from patients with HAM/TSP contained more IFN-γ-expressing cells and fewer interleukin 4-expressing cells than those from healthy donors. Notably, Tax-specific cytotoxic T lymphocytes that may help control the HTLV-1 infection were overwhelmingly CCR4(-). We determined that CD8(+)CCR4(+) T cells and CD4(+)CCR4(+) T cells are prime therapeutic targets for treating HAM/TSP and propose mogamulizumab as a new treatment.
Product Description
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Prednisolone, VETRANAL®, analytical standard
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Prednisolone, ≥99%
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Prednisolone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Prednisolone, United States Pharmacopeia (USP) Reference Standard
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Prednisolone, British Pharmacopoeia (BP) Assay Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O