MilliporeSigma
  • Inactivation of protein-phosphatase 2A causing hyperphosphorylation of autoantigenic paraprotein targets in MGUS/MM is due to an exchange of its regulatory subunits.
Inactivation of protein-phosphatase 2A causing hyperphosphorylation of autoantigenic paraprotein targets in MGUS/MM is due to an exchange of its regulatory subunits.
International journal of cancer (2014-03-29)
Klaus-Dieter Preuss, Natalie Fadle, Evi Regitz, Gerhard Held, Michael Pfreundschuh
ABSTRACT
Hyperphosphorylated paratarg-7 (pP-7) carrier state is the strongest molecularly defined risk factor for monoclonal gammopathy of undetermined significance (MGUS), multiple myeloma (MM) and Waldenstrom's macroglobulinemia (WM). pP-7 is inherited as autosomal-dominant trait and depending on the ethnic background is found in over one-third of MGUS/MM patients. P-7, which is the antigenic paraprotein target in these patients, is hyperphosphorylated at serine17. P-7 hyperphosphorylation can be induced in wild-type P-7 (wtP-7) carriers by PKCζ and reverted by protein-phosphatase 2A (PP2A). Here we show that dephosphorylation of pP-7 is defective in pP-7 carriers due to inactivation of the PP2A by substitution of the regulatory B55δ subunit with B56γ3. In lymphoblastoid cell lines from pP-7 carriers, transfection of recombinant B55δ or treatment with ceramide led to a partial reconstitution of PP2A activity and dephosphorylation of pP-7 to wtP7. Similar results were observed with other previously reported autoantigenic paraproteins targets. In conclusion, the mechanisms responsible for the defective dephosphorylation and maintaining the hyperphosphorylated state of P-7 and other autoantigenic paraprotein targets have been elucidated, facilitating the identification of the genetic basis underlying this phenomenon which is obviously common in the pathogenesis of MGUS/MM/WM and not restricted to pP-7 cases.
Sigma-Aldrich
Anti-methyl-PP2A Antibody, C subunit, clone 2A10, clone 2A10, from mouse