MilliporeSigma
  • Cell wall integrity, genotoxic injury and PCD dynamics in alfalfa saponin-treated white poplar cells highlight a complex link between molecule structure and activity.
Cell wall integrity, genotoxic injury and PCD dynamics in alfalfa saponin-treated white poplar cells highlight a complex link between molecule structure and activity.
Phytochemistry (2015-02-11)
Stefania Paparella, Aldo Tava, Pinarosa Avato, Elisa Biazzi, Anca Macovei, Marco Biggiogera, Daniela Carbonera, Alma Balestrazzi
ABSTRACT
In the present work, eleven saponins and three sapogenins purified from Medicago sativa were tested for their cytotoxicity against highly proliferating white poplar (Populus alba L.) cell suspension cultures. After preliminary screening, four saponins with different structural features in terms of aglycone moieties and sugar chains (saponin 3, a bidesmoside of hederagenin; saponins 4 and 5, monodesmoside and bidesmoside of medicagenic acid respectively, and saponin 10, a bidesmoside of zanhic acid) and different cytotoxicity were selected and used for further investigation on their structure-activity relationship. Transmission Electron Microscopy (TEM) analyses provided for the first time evidence of the effects exerted by saponins on plant cell wall integrity. Exposure to saponin 3 and saponin 10 resulted into disorganization of the outer wall layer and the effect was even more pronounced in white poplar cells treated with the two medicagenic acid derivatives, saponins 4 and 5. Oxidative burst and nitric oxide accumulation were common hallmarks of the response of white poplar cells to saponins. When DNA damage accumulation and DNA repair profiles were evaluated by Single Cell Gel Electrophoresis, induction of single and double strand breaks followed by effective repair was observed within 24h. The reported data are discussed in view of the current issues dealing with saponin structure-activity relationship.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
DAPI, for nucleic acid staining