MilliporeSigma
  • Prospective associations between persistent organic pollutants and metabolic syndrome: a nested case-control study.
Prospective associations between persistent organic pollutants and metabolic syndrome: a nested case-control study.
The Science of the total environment (2014-08-05)
Yu-Mi Lee, Ki-Su Kim, Se-A Kim, Nam-Soo Hong, Su-Jin Lee, Duk-Hee Lee
ABSTRACT
Exposure to persistent organic pollutants (POPs) has recently been linked to metabolic syndrome (MetS) and some MetS components. However, prospective evidence in humans is scarce, and the nature of the dose-response relationship is unclear. We evaluated the association between POPs and MetS using a nested-case control study within a community-based Korean cohort. The study subjects were 64 patients newly diagnosed with MetS during a 4-year follow-up, and the controls were 182 subjects without MetS. Concentrations of polychlorinated biphenyls (PCBs) and organochlorine pesticides (OCPs) were measured in stored serum collected at baseline. The concentrations of most PCBs and some OCPs such as β-hexachlorocyclohexane, hexachlorobenzene, oxychlordane, and heptachlor epoxide predicted the risk for MetS. The POP exposure and MetS showed an inverted U-shaped or a linear association with plateau rather than a linear dose-response association. When the summary measure of the PCBs and OCPs was used, the adjusted odds ratios (ORs) across the quartiles of the summary measure were 1.0, 1.3, 3.8 (95% confidence interval, 1.3-10.7), and 2.1 (Pquadratic=0.013) after adjusting for potential confounders. In the analyses of each of the five MetS components, POP exposure was mainly associated with an increased risk for glucose and lipid metabolism disturbances. This study demonstrated that chronic exposure to a mixture of PCBs and OCPs can increase the risk for MetS within the low-dose background exposure range of POPs. As the findings of this study suggest a nonmonotonic dose-response relationship, in vitro and in vivo experimental studies are needed to understand the underlying mechanisms.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®