MilliporeSigma
  • Intradermal administration of the Type II heat-labile enterotoxins LT-IIb and LT-IIc of enterotoxigenic Escherichia coli enhances humoral and CD8+ T cell immunity to a co-administered antigen.
Intradermal administration of the Type II heat-labile enterotoxins LT-IIb and LT-IIc of enterotoxigenic Escherichia coli enhances humoral and CD8+ T cell immunity to a co-administered antigen.
PloS one (2014-12-24)
John C Hu, Camila Mathias-Santos, Christopher J Greene, Natalie D King-Lyons, Juliana F Rodrigues, George Hajishengallis, Luís C S Ferreira, Terry D Connell
ABSTRACT
Vaccinations are extremely effective at combating infectious diseases. Many conserved antigen (Ag) targets, however, are poorly immunogenic. Protein subunit vaccines frequently elicit only humoral immune responses and fail to confer protection against serious intracellular pathogens. These barriers to vaccine development are often overcome by the use of appropriate adjuvants. Heat-labile enterotoxins (HLT) produced by enterotoxigenic strains of Escherichia coli are potent adjuvants when administered by mucosal or systemic routes. The efficacy of the type II HLT, however, has not been well-defined when administered by the intradermal (ID) route. Using a murine ID immunization model, the adjuvant properties of LT-IIb and LT-IIc, two type II HLTs, were compared with those of LT-I, a prototypical type I HLT. While all three HLT adjuvants enhanced Ag-specific humoral responses to similar levels, LT-IIb and LT-IIc, in contrast to LT-I, induced a more vigorous Ag-specific CD8+ T cell response and proffered faster clearance of Listeria monocytogenes in a challenge model. Additionally, LT-IIb and LT-IIc induced distinct differences in the profiles of the Ag-specific CD8+ T cell responses. While LT-IIc stimulated a robust and rapid primary CD8+ T cell response, LT-IIb exhibited slower CD8+ T cell expansion and contraction kinetics with the formation of higher percentages of effector memory cells. In comparison to LT-I and LT-IIc, LT-IIb evoked better long-term protection after immunization. Furthermore, LT-IIb and LT-IIc enhanced the total number of dendritic cells (DC) in the draining lymph node (DLN) and expression of costimulatory molecules CD80, CD86, and CD40 on DCs. In contrast to LT-I, LT-IIb and LT-IIc induced less edema, cellular infiltrates, and general inflammation at the site of ID injection. Thus, LT-IIb and LT-IIc are attractive comprehensive ID adjuvants with unique characteristic that enhance humoral and cellular immunity to a co-administered protein Ag.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Galactose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material