MilliporeSigma
  • Generation of basic centers in high-silica zeolites and their application in gas-phase upgrading of bio-oil.
Generation of basic centers in high-silica zeolites and their application in gas-phase upgrading of bio-oil.
ChemSusChem (2014-04-24)
Tobias C Keller, Elodie G Rodrigues, Javier Pérez-Ramírez
ABSTRACT
High-silica zeolites have been reported recently as efficient catalysts for liquid- and gas-phase condensation reactions because of the presence of a complementary source of basicity compared to Al-rich basic zeolites. Herein, we describe the controlled generation of these active sites on silica-rich FAU, BEA, and MFI zeolites. Through the application of a mild base treatment in aqueous Na2CO3, alkali-metal-coordinating defects are generated within the zeolite whereas the porous properties are fully preserved. The resulting catalysts were applied in the gas-phase condensation of propanal at 673 K as a model reaction for the catalytic upgrading of pyrolysis oil, for which an up to 20-fold increased activity compared to the unmodified zeolites was attained. The moderate basicity of these new sites leads to a coke resistance superior to traditional base catalysts such as CsX and MgO, and comparable activity and excellent selectivity is achieved for the condensation pathways. Through strategic acid and base treatments and the use of magic-angle spinning NMR spectroscopy, the nature of the active sites was investigated, which supports the theory of siloxy sites as basic centers. This contribution represents a key step in the understanding and design of high-silica base catalysts for the intermediate deoxygenation of crude bio-oil prior to the hydrotreating step for the production of second-generation biofuels.
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)