MilliporeSigma
  • Study of in vitro metabolism of m-nisoldipine in human liver microsomes and recombinant cytochrome P450 enzymes by liquid chromatography-mass spectrometry.
Study of in vitro metabolism of m-nisoldipine in human liver microsomes and recombinant cytochrome P450 enzymes by liquid chromatography-mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2014-05-13)
Lin Yuan, Peipei Jia, Yupeng Sun, Chengcheng Zhao, Xuran Zhi, Ning Sheng, Lantong Zhang
ABSTRACT
This is a report about the investigation of the metabolic fate of m-nisoldipine in human liver microsomes and the recombinant cytochrome P450 enzymes by using LC-MS/MS. A sensitive and reliable LC-MS/MS method was developed to obtain a rapid and complete characterization of new metabolites and the metabolism pathways. The analytes were separated on a reversed phase C18 column with acetonitrile and 0.1% aqueous formic acid as the mobile phase. Tandem mass spectrometry with positive electrospray ionization was used to enable the structural characterization of the metabolites. A total of 10 metabolites were characterized with proposed structures in the incubation of human liver microsomes by comparing their retention times and spectral patterns with those of the parent drug. Dehydrogenation of the dihydropyridine core and reactions of side chains such as hydroxylation and hydrolysis of ester bonds were the major metabolic pathways. The specific cytochrome P450 (CYP) enzymes responsible for m-nisoldipine metabolites were identified using chemical inhibition and cDNA expressed CYP enzymes. The results indicated that CYP2C19 and CYP3A4 might play major roles in the metabolism of m-nisoldipine in human liver microsomes.
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Quinidine, anhydrous