MilliporeSigma
  • Oxidation and phosphorylation of MAP kinase 4 cause protein aggregation.
Oxidation and phosphorylation of MAP kinase 4 cause protein aggregation.
Biochimica et biophysica acta (2014-11-30)
Tong Zhang, Mengmeng Zhu, Wen-yuan Song, Alice C Harmon, Sixue Chen
ABSTRACT
Mitogen-activated protein kinase (MPK) cascades are highly conserved signaling pathways that respond to environmental cues. Arabidopsis MPK4 has been identified as a stress-responsive protein kinase. Here we demonstrate that Brassica napus MPK4 (BnMPK4) is activated by hydrogen peroxide (H2O2) and phytohormone abscisic acid (ABA). Transient expression of a constitutively active BnMPK4 causes H2O2 production and cell death in Nicotiana benthamiana leaves. However, little is known about how H2O2 contributes to the regulation of MPK4 kinase function. Biochemical analysis revealed that recombinant BnMPK4 autophosphorylates on both threonine and tyrosine residues in the activation loop. In the presence of H2O2, phosphorylation of BnMPK4 caused protein aggregation in vitro. The aggregation of BnMPK4 could be reversed to the monomeric form by reducing reagents. Point-mutation of cysteine codons indicated that cysteine 232 is involved in protein aggregation. Our results suggest that BnMPK4 is involved in reactive oxygen species (ROS) signaling and metabolism, and its aggregation may be modulated by redox.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis