MilliporeSigma
  • Emodin inhibits tonic tension through suppressing PKCδ-mediated inhibition of myosin phosphatase in rat isolated thoracic aorta.
Emodin inhibits tonic tension through suppressing PKCδ-mediated inhibition of myosin phosphatase in rat isolated thoracic aorta.
British journal of pharmacology (2014-06-10)
Kyung-Min Lim, Jae-Hyuk Kwon, Keunyoung Kim, Ji-Yoon Noh, Seojin Kang, Jung-Min Park, Moo-Yeol Lee, Ok-Nam Bae, Jin-Ho Chung
ABSTRACT
Dysregulated tonic tension and calcium sensitization in blood vessels has frequently been observed in many cardiovascular diseases. Despite a huge therapeutic potential, little is known about natural products targeting tonic tension and calcium sensitization. We screened natural products for inhibitory effects on vasoconstriction using the rat isolated thoracic aorta and found that an anthraquinone derivative, emodin, attenuated tonic tension. Organ bath system, primary vascular smooth muscle cells, confocal microscopy and Western blot analysis were employed to demonstrate the suppressive effects of emodin on PKCδ-mediated myosin phosphatase inhibition. Emodin, an active ingredient of Polygonum multiflorum extract, inhibited phenylephrine-induced vasoconstriction in rat isolated thoracic aorta, and inhibited vasoconstriction induced by 5-HT and endothelin-1. It also generally suppressed vasoconstrictions mediated by voltage-operated, store-operated calcium channels and intracellular calcium store. However, emodin did not affect agonist-induced calcium increases in primary smooth muscle cells. In contrast, post-treatment with emodin following phenylephrine stimulation potently suppressed tonic tension in rat aortic rings. Western blot analysis revealed that emodin inhibited phenylephrine-induced phospho-myosin light chain (pMLC) and the phosphorylation of myosin-targeting subunit and C-kinase-activated protein phosphatase-1 inhibitor (CPI-17). This was mediated by selective inhibition of PKCδ, whereas PKCα was not involved. Emodin attenuates tonic tension through the blockade of PKCδ and CPI-17-mediated MLC-phosphatase inhibition. This new mode of action for the suppression of tonic tension and structural insights into PKCδ inhibition revealed by emodin may provide new information for the development of modulators of tonic tension and for the treatment of hypertension.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)