MilliporeSigma
  • 1,3-disubstituted-4-aminopyrazolo [3, 4-d] pyrimidines, a new class of potent inhibitors for phospholipase D.
1,3-disubstituted-4-aminopyrazolo [3, 4-d] pyrimidines, a new class of potent inhibitors for phospholipase D.
Chemical biology & drug design (2014-03-20)
Aditya Kulkarni, Phong Quang, Victoriana Curry, Renee Keyes, Weihong Zhou, Hyejin Cho, Jonathan Baffoe, Béla Török, Kimberly Stieglitz
ABSTRACT
Phospholipase D enzymes cleave lipid substrates to produce phosphatidic acid, an important precursor for many essential cellular molecules. Phospholipase D is a target to modulate cancer-cell invasiveness. This study reports synthesis of a new class of phospholipase D inhibitors based on 1,3-disubstituted-4-amino-pyrazolopyrimidine core structure. These molecules were synthesized and used to perform initial screening for the inhibition of purified bacterial phospholipase D, which is highly homologous to the human PLD1 . Initially tested with the bacterial phospholipase D enzyme, then confirmed with the recombinant human PLD1 and PLD2 enzymes, the molecules presented here exhibited inhibition of phospholipase D activity (IC50 ) in the low-nanomolar to low-micromolar range with both monomeric substrate diC4 PC and phospholipid vesicles and micelles. The data strongly indicate that these inhibitory molecules directly block enzyme/vesicle substrate binding. Preliminary activity studies using recombinant human phospholipase Ds in in vivo cell assays measuring both transphosphatidylation and head-group cleavage indicate inhibition in the mid- to low-nanomolar range for these potent inhibitory novel molecules in a physiological environment.
Product Description
Avanti
VU0155056, Avanti Research - A Croda Brand 857370P, powder