MilliporeSigma
  • Chronic activation of sigma-1 receptor evokes nociceptive activation of trigeminal nucleus caudalis in rats.
Chronic activation of sigma-1 receptor evokes nociceptive activation of trigeminal nucleus caudalis in rats.
Pharmacology, biochemistry, and behavior (2014-07-06)
Kihyun Pyun, Ji Seon Son, Young Bae Kwon
ABSTRACT
Primary headache disorders, including migraine, are thought to be mediated by prolonged nociceptive activation of the trigeminal nucleus caudalis (TNC), but the precise mechanisms are poorly understood. Our past studies demonstrated that sigma-1 receptors (Sig-1R) facilitate spinal nociceptive transmission in several pain models. Based on these findings, this study asked if chronic activation of Sig-1R by intracisternal administration of the selective Sig-1R agonist, PRE084, produced TNC neuronal activation as a migraine trigger in rats. A single infusion of PRE084 (10, 50, 100, 500 nmol) significantly increased the number of Fos immunoreactive neurons (Fos-IR) in TNC, which BD1047 (a Sig-1R antagonist) reversed. Chronic infusion of PRE084 (100 nmol for 1, 3, 7 and 14 days) time-dependently elevated Fos-IR in TNC. The number of Fos-IR elevation from day 7 of infusion was comparable with a single capsaicin infusion as a headache model. Increase in face grooming/scratching behavior was evident from day 7, and peaked at day 14 of chronic PRE084 infusion, which was correlated with ΔFosB elevation and phosphorylation of extracellular signal-regulated kinase, and the NMDA receptor NR1 subunit in TNC. Following 14 days of PRE084 infusion, the number of Fos-IR increased until day 7 after final infusion. Moreover, by day 14, Fos-IR associated with PRE084 infusion was significantly reversed by NMDA receptor antagonist MK801, rather than BD1047. These findings indicated that chronic activation of Sig-1R could evoke prolonged neuronal activation in the trigeminovascular system.
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Pricing and availability is not currently available.
Supelco
Capsaicin, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Capsaicin, analytical standard
Pricing and availability is not currently available.
Capsaicin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Capsaicin, natural
Pricing and availability is not currently available.
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Pricing and availability is not currently available.
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.