MilliporeSigma
  • FTY720 does not protect from traumatic brain injury in mice despite reducing posttraumatic inflammation.
FTY720 does not protect from traumatic brain injury in mice despite reducing posttraumatic inflammation.
Journal of neuroimmunology (2014-08-02)
Stine Mencl, Nelli Hennig, Sarah Hopp, Michael K Schuhmann, Christiane Albert-Weissenberger, Anna-Leena Sirén, Christoph Kleinschnitz
ABSTRACT
Inflammation is a pathological hallmark of traumatic brain injury (TBI). Recent evidence suggests that immune cells such as lymphocytes are of particular relevance for lesion development after TBI. FTY720, a sphingosine-1-phosphate (S1P) receptor modulator, sequesters T lymphocytes in lymphoid organs and has been shown to improve outcome in a variety of neurological disease models. We investigated the mode of FTY720 action in models of TBI. Focal cortical cryolesion was induced in C57BL/6 mice treated with FTY720 (1mg/kg) or vehicle immediately before injury. Lesion size was assessed 24h later. Immune cells in the blood and brain were counted by flow cytometry and immunocytochemistry. The integrity of the blood-brain barrier was analyzed using Evans Blue dye. To validate the findings in a diffuse brain trauma model, FTY720-treated mice and controls were subjected to weight drop contusion injury and neurological deficits were assessed until day 7. As expected FTY720 significantly lowered the numbers of circulating lymphocytes and attenuated the invasion of immune cells into the damaged brain parenchyma. However, FTY720 was unable to improve lesion size or functional outcome in both trauma models at either stage, i.e. acute vs chronic. Accordingly, the extent of blood-brain barrier disruption and neuronal apoptosis was similar between FTY720-treated mice and controls. We conclude that pharmacological S1P receptor modulation is an unfavorable strategy to combat TBI. Moreover, our findings put into perspective the pathophysiological relevance of inflammatory cells in traumatic neurodegeneration.
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.