MilliporeSigma
  • Enduring deficits in brain reward function after chronic social defeat in rats: susceptibility, resilience, and antidepressant response.
Enduring deficits in brain reward function after chronic social defeat in rats: susceptibility, resilience, and antidepressant response.
Biological psychiatry (2014-03-01)
Andre Der-Avakian, Michelle S Mazei-Robison, James P Kesby, Eric J Nestler, Athina Markou
ABSTRACT
Anhedonia, or diminished interest or pleasure in rewarding activities, characterizes depression and reflects deficits in brain reward circuitries. Social stress induces anhedonia and increases risk of depression, although the effect of social stress on brain reward function is incompletely understood. This study assessed the following: 1) brain reward function in rats (using the intracranial self-stimulation procedure) and protein levels of brain-derived neurotrophic factor and related signaling molecules in response to chronic social defeat, 2) brain reward function during social defeat and long-term treatment with the antidepressants fluoxetine (5 mg/kg/day) and desipramine (10 mg/kg/day), and 3) forced swim test behavior after social defeat and fluoxetine treatment. Social defeat profoundly and persistently decreased brain reward function, reflecting an enduring anhedonic response, in susceptible rats, whereas resilient rats showed no long-term brain reward deficits. In the ventral tegmental area, social defeat, regardless of susceptibility or resilience, decreased brain-derived neurotrophic factor and increased phosphorylated AKT, whereas only susceptibility was associated with increased phosphorylated mammalian target of rapamycin. Fluoxetine and desipramine reversed lower, but not higher, stress-induced brain reward deficits in susceptible rats. Fluoxetine decreased immobility in the forced swim test, as did social defeat. These results suggest that the differential persistent anhedonic response to psychosocial stress may be mediated by ventral tegmental area signaling molecules independent of brain-derived neurotrophic factor and indicate that greater stress-induced anhedonia is associated with resistance to antidepressant treatment. Consideration of these behavioral and neurobiological factors associated with resistance to stress and antidepressant action may promote the discovery of novel targets to treat stress-related mood disorders.
Product Description
USP
Fluoxetine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Desipramine hydrochloride, ≥98% (TLC), powder
Supelco
Fluoxetine hydrochloride, VETRANAL®, analytical standard
Desipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluoxetine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluoxetine hydrochloride, solid
USP
Desipramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Desipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®