MilliporeSigma
  • p22phox confers resistance to cisplatin, by blocking its entry into the nucleus.
p22phox confers resistance to cisplatin, by blocking its entry into the nucleus.
Oncotarget (2015-02-18)
Chih-Chang Hung, Chen-Yu Chien, Wei-Fan Chiang, Chang-Shen Lin, Tzyh-Chyuan Hour, Hau-Ren Chen, Ling-Feng Wang, Jenq-Yuh Ko, Chi-Hua Chang, Jeff Yi-Fu Chen
ABSTRACT
Cisplatin (CDDP) is a potent chemotherapeutic agent but resistance to the drug remains a major challenge in cancer treatment. To evaluate the efficacy of CDDP in oral squamous cell carcinoma (OSCC), we found that p22phox was highly expressed in CDDP-resistant OSCC specimens. Knockdown of p22phox sensitized OSCC cell lines to CDDP (P < 0.05). Stable overexpression of p22phox augmented CDDP resistance, as evidenced by the significantly higher IC50 values. This cytoprotective effect was attributed to the abrogation of CDDP-induced apoptosis. Akt phosphorylation was increased in p22phox stable lines. However, blocking PI3K/Akt pathway only partially restored CDDP-induced apoptosis. In addition, the overexpressed p22phox in OSCC cells exhibited cytoplasmic localization with enhanced perinuclear expression, consistent with the localization pattern in OSCC specimens. Remarkably, CDDP entry into the nucleus was severely impaired in p22phox-overexpressing cells (P < 0.001), and cytoplasmically accumulated CDDP was co-localized with overexpressed p22phox. This was supported by decreased CDDP-DNA adduct formation and delayed chk1-p53 signaling activation. Together, overexpression of p22phox sequestered CDDP and caused defective CDDP entry into the nucleus, significantly attenuating CDDP-induced apoptosis. Such diminished apoptosis was further abolished by p22phox-activating PI3K/Akt pathway. Our work has suggested a novel biomarker and insight into the mechanism of CDDP resistance.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akt1
Sigma-Aldrich
MISSION® esiRNA, targeting human AKT1, RP11-982M15.2
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cyba
Supelco
Cyclohexanone, Selectophore, ≥99.5%
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Cyclohexanone, 99.8%