MilliporeSigma
  • Prompt bone marrow-derived mesenchymal stem cell therapy enables early porcine heart function recovery from acute myocardial infarction.
Prompt bone marrow-derived mesenchymal stem cell therapy enables early porcine heart function recovery from acute myocardial infarction.
International heart journal (2014-06-27)
Chang-Qing Fan, Steve Leu, Jiunn-Jye Sheu, Yen-Yi Zhen, Tzu-Hsien Tsai, Yung-Lung Chen, Sheng-Ying Chung, Han-Tan Chai, Cheuk-Kwan Sun, Jenq-Lin Yang, Hsueh-Wen Chang, Sheung-Fat Ko, Hon-Kan Yip
ABSTRACT
Impact of early bone marrow-derived mesenchymal stem cell (BMDMSC) implantation on left ventricular (LV) function after AMI was studied.Twelve mini-pigs were equally divided into placebo (AMI through left coronary artery ligation) and cell-treated groups [BMDMSCs (3.0 × 10(7)) implanted into infarct area (IA)] with myocardium harvested by post-AMI day 90. Six healthy animals served as controls.On post-AMI day 90, magnetic resonance imaging showed a lower LV ejection fraction but higher LV dimensions in the placebo group (P < 0.003) that also had increased IAs but reduced wall thickness (P < 0.005). Pro-apoptotic gene expressions (Bax, caspase-3) and apoptotic nucleus number in IAs and peri-IAs were highest in the placebo group (P < 0.001). Inflammatory biomarker expressions (MMP-9, oxidized protein, CD40+ cells) were highest, whereas those of angiogenesis (VEGF, CD31+ cells, SDF-1α, CXCR4) and myocardium-preservation (connexin43, troponin-I, cytochrome-C) were lowest in the placebo group (P < 0.01).BMDMSC implantation preserved LV function and alleviated remodeling at post-AMI day 90.
Sigma-Aldrich
Monoclonal Anti-GJA1 antibody produced in mouse, clone 3E5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.