MilliporeSigma
  • Synergistic geometric and electronic effects for electrochemical reduction of carbon dioxide using gold-copper bimetallic nanoparticles.
Synergistic geometric and electronic effects for electrochemical reduction of carbon dioxide using gold-copper bimetallic nanoparticles.
Nature communications (2014-09-12)
Dohyung Kim, Joaquin Resasco, Yi Yu, Abdullah Mohamed Asiri, Peidong Yang
ABSTRACT
Highly efficient and selective electrochemical reduction of carbon dioxide represents one of the biggest scientific challenges in artificial photosynthesis, where carbon dioxide and water are converted into chemical fuels from solar energy. However, our fundamental understanding of the reaction is still limited and we do not have the capability to design an outstanding catalyst with great activity and selectivity a priori. Here we assemble uniform gold-copper bimetallic nanoparticles with different compositions into ordered monolayers, which serve as a well-defined platform to understand their fundamental catalytic activity in carbon dioxide reduction. We find that two important factors related to intermediate binding, the electronic effect and the geometric effect, dictate the activity of gold-copper bimetallic nanoparticles. These nanoparticle monolayers also show great mass activities, outperforming conventional carbon dioxide reduction catalysts. The insights gained through this study may serve as a foundation for designing better carbon dioxide electrochemical reduction catalysts.
Product Description
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, natural, FCC
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
1,2-Hexadecanediol, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Copper(II) acetate, powder, 99.99% trace metals basis
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Copper(II) acetate, 98%
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
Tetradecylphosphonic acid, 98%