MilliporeSigma
  • B7-H3 promotes aggression and invasion of hepatocellular carcinoma by targeting epithelial-to-mesenchymal transition via JAK2/STAT3/Slug signaling pathway.
B7-H3 promotes aggression and invasion of hepatocellular carcinoma by targeting epithelial-to-mesenchymal transition via JAK2/STAT3/Slug signaling pathway.
Cancer cell international (2015-04-25)
Fu-Biao Kang, Ling Wang, Heng-Chuan Jia, Dong Li, Hai-Jun Li, Yin-Ge Zhang, Dian-Xing Sun
ABSTRACT
B7-homologue 3 (B7-H3), a recently identified immunoregulatory protein, has been shown to be overexpressed in human hepatocellular carcinoma (HCC). However, whether the dynamic expression pattern of B7-H3 contributes to early invasion of HCC is largely unknown. In addition, the biological roles of B7-H3 in HCC are still unclear. Herein, we are going to examine B7-H3 expression profile and its clinicopathological significance in primary and metastatic HCC, and further determine whether B7-H3 knockdown simulates different pathological states of HCC progression and metastasis. Using immunohistochemistry, B7-H3 expression was studied on 116 HCC containing primary and metastatic HCCs. Survival curves and log-rank tests were used to test the association of B7-H3 expression with survival. HCC cells with B7-H3 depletion were established by RNA interference to investigate the effect of B7-H3 on cell proliferation, apoptosis, migration and invasion in vitro. Statistical analysis of clinical cases revealed that B7-H3 high expression group had inclinations towards late TNM stage, the presence of vascular invasion, lymph metastasis, and the formation of microsatellite tumors. Increased intensity of tumor B7-H3 staining was detected more significantly in metastatic HCC tumors. Consistently in experiments performed in vitro, B7-H3 was able to stimulate the wound healing, metastasis and invasion of hepatoma cells by targeting epithelial-to-mesenchymal transition (EMT) via JAK2/Stat3/Slug signaling pathway, while no obvious influence on cell growth and apoptosis. B7-H3 in the regulation of the metastatic capacity of HCC cells makes itself a promising therapeutic target for anti-metastasis therapy.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cd276
Sigma-Aldrich
MISSION® esiRNA, targeting human CD276
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%