MilliporeSigma
  • Blood thrombogenicity is independently associated with serum TSH levels in post-non-ST elevation acute coronary syndrome.
Blood thrombogenicity is independently associated with serum TSH levels in post-non-ST elevation acute coronary syndrome.
The Journal of clinical endocrinology and metabolism (2014-03-19)
Girish Viswanathan, Karthik Balasubramaniam, Richard Hardy, Sally Marshall, Azfar Zaman, Salman Razvi
ABSTRACT
Higher serum TSH levels, both within the reference range and in those with subclinical hypothyroidism (SCH), have been associated with increased risk of atherosclerosis and cardiovascular (CV) events in a number of cross-sectional and longitudinal studies. Our objective was to evaluate blood thrombogenicity in patients post-non-ST elevation acute coronary syndrome (NSTE-ACS) in relation to their thyroid function. DESIGN, PATIENTS, AND OUTCOME MEASURE: At 1 week after troponin-positive NSTE-ACS, 70 patients who had been treated with optimal antiplatelet and secondary prevention therapy were studied. Patients with known thyroid disease or on medications affecting thyroid function were excluded. Blood thrombogenicity was assessed using the ex vivo Badimon perfusion chamber. Serum TSH was associated with higher thrombus burden (β = .30; P = .01) independent of other well-established CV risk factors. Patients with SCH (n = 12; 17%) had a higher thrombus burden than euthyroid individuals as evidenced by the area of the thrombus: mean (SD) 23 608 (10 498) vs 16 661 (10 902) μm(2)/mm (P = .02). However, this association was not evident when the analysis was limited to patients with serum TSH within the reference range. In addition, neither serum free T4 nor free T3 had any significant association with thrombus area. Serum TSH levels, particularly in the SCH range, are associated with higher thrombus burden despite optimal recommended secondary prevention therapy after NSTE-ACS. This may explain the higher CV risk seen in SCH patients. Future trials to assess the effect of individualized antithrombotic as well as thyroid hormone replacement therapy to reduce atherothrombotic risk in this population are needed.
Sigma-Aldrich
Mouse Cd40lg / CD40 Ligand ELISA Kit
Sigma-Aldrich
Rat CD40LG / CD40 Ligand ELISA Kit
Sigma-Aldrich
Human CD40L ELISA Kit, for serum, plasma, cell culture supernatant and urine