MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • MEKK3 and survivin expression in cervical cancer: association with clinicopathological factors and prognosis.
MEKK3 and survivin expression in cervical cancer: association with clinicopathological factors and prognosis.
Asian Pacific journal of cancer prevention : APJCP (2014-07-22)
Xue-Quan Cao, Hong-Sheng Lu, Ling Zhang, Li-Li Chen, Mei-Fu Gan
ABSTRACT
Mitogen-activated protein kinase/extracellular signal-regulated kinase kinase kinase 3 (MEKK3) is an important protein kinase and a member of the MAPK family, which regulates cellular responses to environmental stress and serves as key integration points along the signal transduction cascade that not only link diverse extracellular stimuli to subsequent signaling molecules but also amplify the initiating signals to ultimately activate effector molecules and induce cell proliferation, differentiation and survival. To explore the relationship between MEKK3 and cell apoptosis, clinicopathology and prognosis, we characterize the expression of MEKK3 and survivin in cervical cancer. MEKK3 and survivin expression was measured by RT-PCR and Western blotting of fresh surgical resections from 30 cases of cervical cancer and 25 cases of chronic cervicitis. Protein expression was detected by tissue microarray and immunochemistry (En Vision) in 107 cases of cervical cancer, 86 cases of cervical intraepithelial neoplasia (CIN), and 35 cases of chronic cervicitis. Expression patterns were analyzed for their association with clinicopathological factors and prognosis in cervical cancer. Expression of MEKK3 and survivin mRNA was significantly higher in cervical cancer than in the controls (p<0.05). MEKK3 and survivin expression differed significantly between cervical carcinoma, CIN, and cervicitis (p<0.05) and correlated with clinical stage, infiltration depth, and lymph node metastasis (p<0.05). MEKK3 expression was positively correlated with survivin (p<0.05). Kaplan-Meier survival analysis showed that MEKK3 and survivin expression, lymph node metastasis, depth of invasion, and FIGO stage reduce cumulative survival. Cox multivariate regression analysis showed that MEKK3, survivin, and clinical staging are independent prognostic factors in cervical cancer (p<0.05). Expression of MEKK3 and survivin are significantly increased in cervical cancer, their overexpression participating in the occurrence and development of cervical cancer, with protein expression and clinical staging acting as independent prognostic factors for patients with cervical cancer.