MilliporeSigma
A modular, air-stable nickel precatalyst.
Organic letters (2015-04-18)
Jason D Shields, Erin E Gray, Abigail G Doyle
ABSTRACT
The synthesis and catalytic activity of [(TMEDA)Ni(o-tolyl)Cl], an air-stable, crystalline solid, is described. This complex is an effective precatalyst in a variety of nickel-catalyzed transformations. The lability of TMEDA allows a wide variety of ligands to be used, including mono- and bidentate phosphines, diimines, and N-heterocyclic carbenes. Preliminary mechanistic studies are also reported, which suggest that [(TMEDA)Ni(o-tolyl)Cl] can activate by either a Ni-B or Ni-Ni transmetalation event, depending on the reaction conditions.
Sigma-Aldrich
[(TMEDA)Ni(o-tolyl)Cl], 95%