MilliporeSigma
  • Pharmacodynamic and pharmacokinetic characterization of poly(ethylene glycol) conjugation to met-enkephalin analog [D-Pen2, D-Pen5]-enkephalin (DPDPE).
Pharmacodynamic and pharmacokinetic characterization of poly(ethylene glycol) conjugation to met-enkephalin analog [D-Pen2, D-Pen5]-enkephalin (DPDPE).
The Journal of pharmacology and experimental therapeutics (2001-07-17)
K A Witt, J D Huber, R D Egleton, M J Roberts, M D Bentley, L Guo, H Wei, H I Yamamura, T P Davis
ABSTRACT
Poly(ethylene glycol), or PEG, conjugation to proteins and peptides is a growing technology used to enhance efficacy of therapeutics. This investigation assesses pharmacodynamic and pharmacokinetic characteristics of PEG-conjugated [D-Pen2,D-Pen5]-enkephalin (DPDPE), a met-enkephalin analog, in rodent (in vivo, in situ) and bovine (in vitro) systems. PEG-DPDPE showed increased analgesia (i.v.) compared with nonconjugated form (p < 0.01), despite a 172-fold lower binding affinity for the delta-opioid receptor. [125I]PEG-DPDPE had a 36-fold greater hydrophilicity (p < 0.01) and 12% increase in the unbound plasma protein fraction (p < 0.01), compared with [(125)I]DPDPE. [125I]PEG-DPDPE had a 2.5-fold increase in elimination half-life (p < 0.01), 2.7-fold decrease in volume of distribution (p < 0.01), and a 7-fold decrease in plasma clearance rate (p < 0.01) to [125I]DPDPE. Time course distribution showed significant concentration differences (p < 0.01) in plasma, whole blood, liver, gallbladder, gastrointestinal (GI) content, GI tract, kidneys, spleen, urine, and brain (brain, p < 0.05), between the conjugated and nonconjugated forms. Increased brain uptake of [(125)I]PEG-DPDPE corresponded to analgesia data. [125I]PEG-DPDPE in brain was shown to be 58.9% intact, with 41.1% existing as [125I]DPDPE (metabolite), whereas [125I]DPDPE was 25.7% intact in the brain (at 30 min). In vitro P-glycoprotein affinity was shown for [125I]DPDPE (p < 0.01) but not shown for [125I]PEG-DPDPE. In vitro saturable uptake, with 100 microM DPDPE, was shown for [125I]PEG-DPDPE (p < 0.05). In this study, PEG-conjugated DPDPE seems to act as a prodrug, enhancing peripheral pharmacokinetics, while undergoing hydrolysis in the brain and allowing nonconjugated DPDPE to act at the receptor.
Sigma-Aldrich
[D-Pen2,5]-Enkephalin hydrate, ≥95% (HPLC)