MilliporeSigma
  • DISC1-kendrin interaction is involved in centrosomal microtubule network formation.
DISC1-kendrin interaction is involved in centrosomal microtubule network formation.
Biochemical and biophysical research communications (2008-10-29)
Shoko Shimizu, Shinsuke Matsuzaki, Tsuyoshi Hattori, Natsuko Kumamoto, Ko Miyoshi, Taiichi Katayama, Masaya Tohyama
ABSTRACT
Disrupted-In-Schizophrenia 1 (DISC1) was identified as a novel gene disrupted by a (1;11)(q42.1;q14.3) translocation segregating with schizophrenia, bipolar disorder and other major mental illnesses in a Scottish family. We previously identified 446-533 amino acids of DISC1 as the kendrin-binding region by means of a directed yeast two-hybrid interaction assay and showed that the DISC1-kendrin interaction is indispensable for the centrosomal localization of DISC1. In this study, to confirm the DISC1-kendrin interaction, we examined the interaction between deletion mutants of DISC1 and kendrin. Then, we demonstrated that the carboxy-terminus of DISC1 is indispensable for the interaction with kendrin. Furthermore, the immunocytochemistry revealed that the carboxy-terminus of DISC1 is also required for the centrosomal targeting of DISC1. Overexpression of the DISC1-binding region of kendrin or the DISC1 deletion mutant lacking the kendrin-binding region impairs the microtubule organization. These findings suggest that the DISC1-kendrin interaction plays a key role in the microtubule dynamics.