MilliporeSigma
  • Diode laser measurements of CD3 quantum yields and internal energy for the dissociation of dimethyl sulfoxide-d6.
Diode laser measurements of CD<SUB>3</SUB> quantum yields and internal energy for the dissociation of dimethyl sulfoxide-d<SUB>6</SUB>.
J. Chem. Phys. (1997)
Rudolph RN, et al.