MilliporeSigma
  • Association of RIP2 gene polymorphisms and systemic lupus erythematosus in a Chinese population.
Association of RIP2 gene polymorphisms and systemic lupus erythematosus in a Chinese population.
Mutagenesis (2011-11-15)
Jing Li, Jing Tian, Yan Ma, Han Cen, Rui-Xue Leng, Man-Man Lu, Gui-Mei Chen, Chen-Chen Feng, Jin-Hui Tao, Hai-Feng Pan, Dong-Qing Ye
ABSTRACT
The aim of this study was to investigate the association of receptor interacting protein 2 (RIP2) single-nucleotide polymorphisms (SNPs) with susceptibility to systemic lupus erythematosus (SLE) in a Chinese population. A case-control study was performed on the SNPs rs16900617 and rs16900627 in 590 Chinese SLE patients and 660 healthy controls. These SNPs were typed by TaqMan allele discrimination assays. We found a significant association of rs16900617 G allele [odds ratio (OR) = 0.54, 95% confidence interval (CI) 0.41-0.72] and rs16900627 G allele (OR = 1.28, 95% CI 1.04-1.58) with SLE. Significant differences in genotype frequency distribution were also found in SLE and control individuals (rs16900617: AG versus AA, OR = 0.59, 95% CI 0.44-0.81; GG versus AA, OR = 0.08, 95% CI 0.01-0.65; AG + GG versus AA, OR = 0.55, 95% CI 0.41-0.75; rs16900627: AG versus AA, OR = 1.51, 95% CI 1.17-1.93; AG + GG versus AA, OR = 1.43, 95% CI 1.13-1.82). Analysis of the haplotypes revealed that two haplotypes of AG and GA were also significantly associated with SLE (OR = 1.37, 95% CI 1.11-1.70; OR = 0.60, 95% CI 0.45-0.79). Our findings suggest that the RIP2 gene polymorphisms may be associated with susceptibility to SLE in the Chinese population.