MilliporeSigma
  • Beta1,4-N-acetylgalactosaminyltransferase (GM2 synthase) is released from Golgi membranes as a neuraminidase-sensitive, disulfide-bonded dimer by a cathepsin D-like protease.
Beta1,4-N-acetylgalactosaminyltransferase (GM2 synthase) is released from Golgi membranes as a neuraminidase-sensitive, disulfide-bonded dimer by a cathepsin D-like protease.
The Journal of biological chemistry (1996-10-18)
E Jaskiewicz, G Zhu, R Bassi, D S Darling, W W Young
ABSTRACT
Many Golgi membrane-bound glycosyltransferases are released from cells in a soluble form. To characterize this release process, we stably transfected Chinese hamster ovary cells with three myc epitope-tagged forms of cloned beta1, 4-N-acetylgalactosaminyltransferase (GalNAcT); two of these forms resided in the Golgi, while the third was retained in the ER. GalNAcT was released into the culture medium from cells transfected with the Golgi forms but not with the ER form of the enzyme. The medium from cells transfected with the Golgi forms contained disulfide-bonded dimers of GalNAcT, which carried neuraminidase sensitive, complex N-linked carbohydrate chains. This soluble species represented the major degradation product of cellular GalNAcT, which turned over with a half-time of about 1.7 h. The soluble species consisted of a mixture of truncated GalNAcT molecules, the major form of which was produced by cleavage near the boundary between the transmembrane and lumenal domains between Leu-23 and Tyr-24. This cleavage site fits the sequence pattern for sites cleaved by cathepsin D (van Noort, J.M., and van der Drift, A. C.M. (1989) J. Biol. Chem. 264, 14159-14164). These findings suggest that GalNAcT is converted from a membrane-bound to a soluble form as a result of cleavage by a cathepsin D-like protease in a compartment late in the Golgi secretory pathway.