MilliporeSigma
  • Clathrin light chains are required for the gyrating-clathrin recycling pathway and thereby promote cell migration.
Clathrin light chains are required for the gyrating-clathrin recycling pathway and thereby promote cell migration.
Nature communications (2014-05-24)
Sophia R Majeed, Lavanya Vasudevan, Chih-Ying Chen, Yi Luo, Jorge A Torres, Timothy M Evans, Andrew Sharkey, Amy B Foraker, Nicole M L Wong, Christopher Esk, Theresa A Freeman, Ashley Moffett, James H Keen, Frances M Brodsky
ABSTRACT
The clathrin light chain (CLC) subunits participate in several membrane traffic pathways involving both clathrin and actin, through binding the actin-organizing huntingtin-interacting proteins (Hip). However, CLCs are dispensable for clathrin-mediated endocytosis of many cargoes. Here we observe that CLC depletion affects cell migration through Hip binding and reduces surface expression of β1-integrin by interference with recycling following normal endocytosis of inactive β1-integrin. CLC depletion and expression of a modified CLC also inhibit the appearance of gyrating (G)-clathrin structures, known mediators of rapid recycling of transferrin receptor from endosomes. Expression of the modified CLC reduces β1-integrin and transferrin receptor recycling, as well as cell migration, implicating G-clathrin in these processes. Supporting a physiological role for CLC in migration, the CLCb isoform of CLC is upregulated in migratory human trophoblast cells during uterine invasion. Together, these studies establish CLCs as mediating clathrin-actin interactions needed for recycling by G-clathrin during migration.
Sigma-Aldrich
Anti-Huntingtin-Interacting Protein 1 Related (HIP1R) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Paxillin Antibody, clone 5H11, clone 5H11, Upstate®, from mouse