MilliporeSigma
Interaction of PC4 with melted DNA inhibits transcription.
The EMBO journal (1998-09-02)
S Werten, G Stelzer, A Goppelt, F M Langen, P Gros, H T Timmers, P C Van der Vliet, M Meisterernst
ABSTRACT
PC4 is a nuclear DNA-binding protein that stimulates activator-dependent class II gene transcription in vitro. Recent biochemical and X-ray analyses have revealed a unique structure within the C-terminal domain of PC4 that binds tightly to unpaired double-stranded (ds)DNA. The cellular function of this evolutionarily conserved dimeric DNA-binding fold is unknown. Here we demonstrate that PC4 represses transcription through this motif. Interaction with melted promoters is not required for activator-dependent transcription in vitro. The inhibitory activity is attenuated on bona fide promoters by (i) transcription factor TFIIH and (ii) phosphorylation of PC4. PC4 remains a potent inhibitor of transcription in regions containing unpaired ds DNA, in single-stranded DNA that can fold into two antiparallel strands, and on DNA ends. Our observations are consistent with a novel inhibitory function of PC4.