MilliporeSigma
  • Intron 3 of the ARID5B gene: a hot spot for acute lymphoblastic leukemia susceptibility.
Intron 3 of the ARID5B gene: a hot spot for acute lymphoblastic leukemia susceptibility.
Journal of cancer research and clinical oncology (2013-09-10)
Ángela Gutiérrez-Camino, Elixabet López-López, Idoia Martín-Guerrero, José Sánchez-Toledo, Nagore García de Andoin, Ana Carboné Bañeres, Purificación García-Miguel, Aurora Navajas, África García-Orad
ABSTRACT
Single-nucleotide polymorphisms (SNPs) in AT-rich interactive domain 5B (ARID5B) have been associated with risk for pediatric acute lymphoblastic leukemia (ALL). After reviewing previous studies, we realized that the most significant associations were restricted to intron 3, but the mechanism(s) by which those SNPs affect ALL risk remain to be elucidated. Therefore, the aim of this study was to analyze the association between genetic variants of the intron 3 region of ARID5B and the incidence of B-ALL in a Spanish population. We also aimed to find a functional explanation for the association, searching for copy number variations (CNVs), and changes in ARID5B expression associated with the genotypes of the SNPs. We analyzed 10 SNPs in intron 3 of ARID5B in a Spanish population of 219 B-ALL patients and 397 unrelated controls with the Taqman Open Array platform. CNVs were analyzed in 23 patients and 17 controls using the Cytogenetics Whole-genome 2.7 M platform. Expression of ARID5B transcript 1 was quantified by qPCR and related to SNPs genotype in seven ALL cell lines. Association between intron 3 and B-ALL risk was confirmed for all of the SNPs evaluated in our Spanish population. We could not explain this association by the presence of CNVs. We neither detected changes in the expression of ARID5B isoform associated with the genotype of the SNPs. The intron 3 of ARID5B gene was found to be strongly associated with B-ALL risk in the Spanish population examined. However, neither CNVs nor changes in mRNA expression were found to be responsible for this association.