MilliporeSigma
  • 2-azaadamantane N-oxyl (AZADO) and 1-Me-AZADO: highly efficient organocatalysts for oxidation of alcohols.
2-azaadamantane N-oxyl (AZADO) and 1-Me-AZADO: highly efficient organocatalysts for oxidation of alcohols.
Journal of the American Chemical Society (2006-06-29)
Masatoshi Shibuya, Masaki Tomizawa, Iwao Suzuki, Yoshiharu Iwabuchi
ABSTRACT
Development of a stable nitroxyl radical class of catalysts, 2-azaadamantane N-oxyl (AZADO) and 1-Me-AZADO, for highly efficient oxidation of alcohols is described. AZADO and 1-Me-AZADO exhibit superior catalytic proficiency to TEMPO, converting various sterically hindered alcohols to the corresponding carbonyl compounds in excellent yields.
Sigma-Aldrich
2-Azaadamantane-N-oxyl, 90%
Pricing and availability is not currently available.