MilliporeSigma
Galactose fermentation and classification of thermophilic lactobacilli.
Applied and environmental microbiology (1983-06-01)
K W Turner, F G Martley
ABSTRACT
The ability to ferment galactose is a major characteristic which can be used to differentiate Lactobacillus helveticus (galactose positive) from Lactobacillus lactis and Lactobacillus bulgaricus (galactose negative). In milk cultures, galactose-positive strains produced d- and l-lactic acid with little galactose accumulation, whereas galactose-negative strains produced d-lactic acid, and galactose accumulated to high levels.
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)