MilliporeSigma
  • Aberrant methylation of PCDH8 is a potential prognostic biomarker for patients with clear cell renal cell carcinoma.
Aberrant methylation of PCDH8 is a potential prognostic biomarker for patients with clear cell renal cell carcinoma.
Medical science monitor : international medical journal of experimental and clinical research (2014-11-25)
Ying-Li Lin, Yan-Ling Wang, Xing-Li Fu, Jian-Guo Ma
ABSTRACT
PCDH8 is a tumor suppressor that regulates cell adhesin, proliferation, and migration. It is often inactivated by aberrant promoter methylation in several human cancers, including clear cell renal cell carcinoma (CCRCC). The clinical significance of PCDH8 methylation in CCRCC remains unclear. The aim of this study was to investigate the relationship between PCDH8 methylation and clinicopathological characteristics as well as outcome of patients with CCRCC. The methylation status of PCDH8 in 153 CCRCC tissues and 97 paired adjacent normal renal tissues were examined using methylation-specific PCR (MSP). Then the relationships between PCDH8 methylation and clinicopathological features as well as progression-free survival of CCRCC patients were evaluated. PCDH8 methylation was significantly more frequent in CCRCC tissues compared with normal renal tissues. Moreover, PCDH8 methylation was significantly correlated with advanced clinical stage (P=0.0141), higher grade (P=0.0190), and lymph node metastasis (P=0.0098). In addition, multivariate analysis showed that PCDH8 methylation was independently associated with poor progression-free survival (P=0.0316). PCDH8 methylation is a frequent event in CCRCC and is correlated with unfavorable clinicopathological features. Moreover, PCDH8 methylation may be a useful biomarker to predict the progression of CCRCC.