MilliporeSigma
  • One-pot synthesis of 3,4-dihydropyrimidin-2(1H)-ones using chloroacetic acid as catalyst.
One-pot synthesis of 3,4-dihydropyrimidin-2(1H)-ones using chloroacetic acid as catalyst.
Bioorganic & medicinal chemistry letters (2007-05-11)
Yang Yu, Di Liu, Chunsheng Liu, Genxiang Luo
ABSTRACT
A simple and effective synthesis of 3,4-dihydropyrimidin-2(1H)-one derivatives from aldehydes, 1,3-dicarbonyl compounds and urea or thiourea using chloroacetic acid as catalyst under solvent-free conditions is described. Compared with the classical Biginelli reaction conditions, this new method has the advantage of good to excellent yields and short reaction time.